Would you like to log in again?
Thermostable RNase H
Supplier: New England Biolabs
M0523S M0523S
76284-680EA 163 USD InStock
76284-680 CA76284-680
Thermostable RNase H
Enzymes
Thermostable RNase H is an endoribonuclease that is functional at high temperatures and selectively hydrolyzes the phosphodiester bonds of an RNA strand in an RNA:DNA hybrid molecule, while leaving the DNA strand intact.
  • Recombinant enzyme supplied with 10X Reaction Buffer
  • Does not digest single or double-stranded DNA
  • Active and stable above 37 °C over a broad temperature range
  • Requires magnesium chloride for activityInactivated with proteinase K treatment or addition of excess EDTA
In addition, Thermostable RNase H does not degrade single- or double-stranded RNA or DNA. While the Thermostable and E. coli RNase H homologs both exhibit these same endoribonucleolytic properties, the thermostable enzyme displays activity at higher temperatures with optimal activity above 65 °C. These properties allow use in applications where higher assay temperatures are desired.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty