Would you like to log in again?
Esp3I
Supplier: New England Biolabs
R0734S R0734S R0734L R0734L
76284-690EA 85 USD InStock
CAMSPP-R0734S 76284-690 76284-688 CA76284-688
Esp3I
Enzymes
A Type IIS restriction endonuclease that recognizes the sequence CGTCTCN^NNNN_....
  • Time-Saver qualified for digestion in 5-15 minutes
  • Isoschizomer of BsmBI-v2
  • Used in Golden Gate AssemblyType IIS REs recognize asymmetric DNA sequences; cleave outside their recognition sequence
  • Supplied with 1 vial of Gel Loading Dye, Purple (6X)
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty