Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-APP Mouse Monoclonal Antibody [clone: WO2]

Supplier: Enzo Life Sciences
ABS386-0100
76284-702EA 635.83 USD InStock
76284-702
Anti-APP Mouse Monoclonal Antibody [clone: WO2]
Antibodies
Recombinant monclonal antibody (Mouse IgG2aκ).

Recommended Applications: ELISA, IHC (PS), WB

Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: WO2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2aκ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty