Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MYC Rat Monoclonal Antibody [clone: 9E10]

Supplier: Enzo Life Sciences
ABS472-0200
76284-806EA 622 USD InStock
76284-806
Anti-MYC Rat Monoclonal Antibody [clone: 9E10]
Antibodies
Recombinant monclonal antibody (Rat IgG2cκ).

Recommended Applications: IF, IHC (FS), IHC (PS), IP, WB

Type: Primary
Antigen: MYC
Clonality: Monoclonal
Clone: 9E10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2cκ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty