Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-His Tag Rabbit Monoclonal Antibody [clone: 3D5]

Supplier: Enzo Life Sciences
ABS528-0200
76284-886EA 622 USD InStock
76284-886
Anti-His Tag Rabbit Monoclonal Antibody [clone: 3D5]
Antibodies
Recombinant monclonal antibody (Rabbit IgGκ).

Recommended Applications: ELISA, WB

Type: Primary
Antigen: His Tag
Clonality: Monoclonal
Clone: 3D5
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgGκ
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty