Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFSF4 Human Monoclonal Antibody [clone: R4930 (Oxelumab)]

Supplier: Enzo Life Sciences
ABS556-0200
76284-900EA 633 USD InStock
76284-900
Anti-TNFSF4 Human Monoclonal Antibody [clone: R4930 (Oxelumab)]
Antibodies
Recombinant monclonal antibody (Human IgG1κ).

Recommended Applications: Flow Cytometry, FUNC

Type: Primary
Antigen: TNFSF4
Clonality: Monoclonal
Clone: R4930 (Oxelumab)
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1κ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty