Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human IgG3 kappa Isotype Control [clone: MOPC-21]

Supplier: Enzo Life Sciences
ABS583-0200
76284-918EA 622 USD InStock
76284-918
Human IgG3 kappa Isotype Control [clone: MOPC-21]
Antibodies
Recombinant monclonal antibody (Human IgG3κ).

Recommended Applications: Flow Cytometry, WB

Type: Control
Antigen:
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG3kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty