Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: Ki-3]

Supplier: Enzo Life Sciences
ABS432-0200
76285-004EA 622 USD InStock
76285-004
Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: Ki-3]
Antibodies
Recombinant monclonal antibody (Mouse IgG2bκ).

Recommended Applications: ELISA, Flow Cytometry, IF, IHC

Type: Primary
Antigen: TNFRSF8
Clonality: Monoclonal
Clone: Ki-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2bκ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty