Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CR1 Rat Monoclonal Antibody [clone: 8C12]

Supplier: Enzo Life Sciences
ABS436-0200
76285-012EA 589.97 USD InStock
76285-012
Anti-CR1 Rat Monoclonal Antibody [clone: 8C12]
Antibodies
Recombinant monclonal antibody (Rat IgG2aκ).

Recommended Applications: Flow Cytometry, IHC (FS), IP, WB

Type: Primary
Antigen: CR1
Clonality: Monoclonal
Clone: 8C12
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2aκ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty