Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-His Tag Human Monoclonal Antibody [clone: 3D5]

Supplier: Enzo Life Sciences
ABS532-0200
76285-056EA 633 USD InStock
76285-056
Anti-His Tag Human Monoclonal Antibody [clone: 3D5]
Antibodies
Recombinant monclonal antibody (Human IgG2κ).

Recommended Applications: ELISA, WB

Type: Primary
Antigen: His Tag
Clonality: Monoclonal
Clone: 3D5
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG2κ
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty