Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-p53 Mouse Monoclonal Antibody [clone: PAb421]

Supplier: Enzo Life Sciences
ABS653-0200
76285-166EA 633 USD InStock
76285-166
Anti-p53 Mouse Monoclonal Antibody [clone: PAb421]
Antibodies
Recombinant monclonal antibody (Mouse IgG2aκ).

Recommended Applications: ELISA, IF, IHC (FS), IHC (PS), IP, WB

Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone: PAb421
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2aκ
Reactivity: Human, Mouse, Rat, Rabbit, Monkey

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty