Would you like to log in again?
Recombinant Albumin, Molecular Biology Grade
Supplier: New England Biolabs
B9200S B9200S
76285-184EA 49 USD InStock
CA76285-184 76285-184
Recombinant Albumin, Molecular Biology Grade
Proteins and Peptides
Recombinant Albumin, Molecular Biology Grade, is a non-bovine derived albumin that can serve as an alternative to Bovine Serum Albumin (BSA).
Like BSA, it has been shown to prevent adhesion of enzymes to reaction tubes and pipette surfaces. It also stabilizes some proteins during incubation. Choose Recombinant Albumin, when there is a need to avoid BSA. Please inquire about custom formulations with Recombinant Albumin.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty