Would you like to log in again?
NEBNext Multiplex Oligos for Illumina (96 Unique Dual Index Primer Pairs), New England Biolabs
Supplier: New England Biolabs
E6440L E6440S E6440S E6440L
76285-192EA 2093 USD InStock
CA76285-192 76285-192 76284-670 CA76284-670
NEBNext Multiplex Oligos for Illumina (96 Unique Dual Index Primer Pairs), New England Biolabs
Nucleic Acid Reagents Primers and Probes
NEBNext Multiplex Oligos provide adaptors and primers to enable high yield multiplex Illumina library production. Unique Dual Index (UDI) primer pairs enable the identification and mitigation of the impacts of index hopping when using a patterened flow cell.
  • Allows complete filtering of index-hopped reads
  • Increased sample identification specificity
  • Minimized adaptor-dimer formation
  • Includes index primers for library multiplexing
  • Combine sets 1 to 4 for 384 barcodes
The unique hairpin loop structure of the NEBNext Adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. This kit includes 96 pre-mixed unique pairs of i5 and i7 index primers, packaged in a single-use 96-well plate with a pierceable foil seal. Additional NEBNext Multiplex Oligos products are available for different indexing strategies.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty