Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VEGFA Rabbit Monoclonal Antibody [clone: Bevacizumab]

Supplier: Enzo Life Sciences
ABS383-0200
76285-224EA 633 USD InStock
76285-224
Anti-VEGFA Rabbit Monoclonal Antibody [clone: Bevacizumab]
Antibodies
Recombinant monclonal antibody (Rabbit IgGκ).

Recommended Applications: ELISA, Flow Cytometry, IP, WB, FUNC

Type: Primary
Antigen: VEGFA
Clonality: Monoclonal
Clone: Bevacizumab
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgGκ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty