Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Star Puffs, Certified

Supplier: Bio-Serv
F7364 F7364
76285-266BG 30.67 USD InStock
CA76285-266 76285-266
Star Puffs, Certified
Animal Supplies Animal Feed
Delicious banana-flavored light crunchy puff.

  • Certified (contaminant screened)
  • A low sugar, low calorie treat (<1 kcal/puff)
  • Made with simple wholesome ingredients
  • Excellent for reward or foraging opportunities
  • Shelf life: 12 months, cool dry storage

Ingredients: Wheat flour, rice flour, wheat middlings, sucrose, banana flakes, apple fiber, lecithin, banana flavoring.

Caloric value: 0.9 kcal/g
Nutritional ash content: 1.3%
Nutritional carbohydrate content: 76%
Nutritional fat content: 3.4%
Nutritional fibre content: 2.6%
Nutritional moisture content: <10%
Nutritional protein content: 7.3%

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty