Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sunflower Seeds, Black Oil, Sterile, Bio-Serv

Supplier: Bio-Serv
S5137 S5137 S5137-1 S5137-1
89067-448EA 79.42 USD InStock
89067-448 CA89067-448 CA76285-298 76285-298
Sunflower Seeds, Black Oil, Sterile, Bio-Serv
Animal Supplies Animal Feed
A healthy, natural foraging treat.

  • Excellent source of omega 6 essential fatty acids, dietary fiber, protein and vitamin E
  • Shelf Life: 12 months, cool dry storage

Caloric value: 4.48 kcal/g
Nutritional ash content: 2.10%
Nutritional carbohydrate content: 0.70%
Nutritional fat content: 41.80%
Nutritional fibre content: 34.20%
Nutritional moisture content: <10%
Nutritional protein content: 17.20%

Ordering information: Due to the method used to process sunflower seeds, this package may contain other plant material.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty