Would you like to log in again?
Fine Tip Transfer Pipettes, Globe Scientific
Supplier: Globe Scientific
134090 134070 134050-S01 134050 134090-S01 134060-S20 134020 134010 134070-S20 134070-S01 134060 134060-S01 134050-S20 134090-S20 134020-S01
76285-414CS 102.1 USD InStock
76285-414 89497-638 89497-637 76285-412 89497-634 76285-408 89497-632 76285-416 89497-642 89497-630 76285-384 89497-640 76285-382 76285-410 76285-380
Fine Tip Transfer Pipettes, Globe Scientific
Pipettes Pasteur/Transfer Pipettes
Produced from a custom grade low-density polyethylene (LDPE), these unbreakable one-piece pipettes are used to draw and dispense liquids in controlled, reproducible drop sizes.
  • Non-toxic and chemically inert to biological fluids and most acids
  • Easy to grip with gloves
  • Temperature Range: –50 to 90 °C
  • Can be gas (EtO) sterilized
Fine tip transfer pipettes are ideal for dispensing small volumes, loading hemocytometers, and transferring liquid to and from multi-well plates.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty