Would you like to log in again?
Non-Graduated Transfer Pipettes, Globe Scientific
Supplier: Globe Scientific
138030 138060-S20 138040 138080-S01 138030-S01 138050 138050-S01 138050-S20 138060 138030-S20 138060-S01 138040-S01 138090 138020-1000 138020-S01 138020B 138080 138040-S20 138090-S01 138080-S20 138020-S20
89497-704CS 297.91 USD InStock
89497-704 490016-614 76285-402 89497-700 89497-698 89497-696 76285-396 76285-374 89497-694 76285-394 76285-372 89497-692 76285-366 76285-400 89497-691 76285-378 76285-364 76285-376 76285-398 76285-392 76285-390
Non-Graduated Transfer Pipettes, Globe Scientific
Pipettes Pasteur/Transfer Pipettes
Produced from a custom grade low-density polyethylene (LDPE), these unbreakable one-piece pipettes are used to draw and dispense liquids in controlled, reproducible drop sizes.
  • Non-toxic and chemically inert to biological fluids and most acids
  • Easy to grip with gloves
  • Temperature Range: –50 to +90 °C
  • Can be gas (EtO) sterilized
Offered in a variety of styles, these popular non-graduated general purpose transfer pipettes are packaged in a unique dispenser box. The convenient box is small, compact, and ideal for benchtop use. The hinged front panel opens for quick access and can be closed when not in use to keep the contents clean. Each dispenser box contains 500 pipettes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty