Would you like to log in again?
Narrow Stem Transfer Pipettes, Globe Scientific
Supplier: Globe Scientific
136030-S20 136030-S01 139040-S20 139040 136020-S20 136020 136030 136020-S01 139020 139040-S01 139040-250
89497-714PK 39.36 USD InStock
89497-714 76285-368 76285-424 89497-712 76285-370 89497-654 89497-652 76285-388 76285-422 76285-386 89497-706
Narrow Stem Transfer Pipettes, Globe Scientific
Pipettes Pasteur/Transfer Pipettes
Produced from a custom grade low-density polyethylene (LDPE), these unbreakable one-piece pipettes are used to draw and dispense liquids in controlled, reproducible drop sizes.
  • Non-toxic and chemically inert to biological fluids and most acids
  • Easy to grip with gloves
  • Temperature Range: –50 to +90 °C
  • Can be gas (EtO) sterilized
Narrow stem transfer pipettes feature a long, thin stem that is flexible and can be bent to access narrow openings and small tube diameters.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty