Would you like to log in again?
Graduated Transfer Pipettes, Globe Scientific
Supplier: Globe Scientific
136036 137040-S01 135038 135010-S01 135010 137030 137238 139060 136036-S01 137040 137135 137040-S20 135238 137138 137050 137035-S20 138070 137010-S20 137035 137135-100 136036-S20 135138 137010
10799-769CS 170.83 USD InStock
10799-769 76285-426 89497-648 76285-434 89497-646 89497-668 89497-656 89497-678 89497-644 89497-666 89497-676 76285-428 89497-674 89497-672 89497-650 76285-360 75845-068 76285-432 89497-670 89497-680 76285-430 76285-420 76247-047
Graduated Transfer Pipettes, Globe Scientific
Pipettes Pasteur/Transfer Pipettes
Produced from a custom grade low-density polyethylene (LDPE), these unbreakable one-piece pipettes are used to draw and dispense liquids in controlled, reproducible drop sizes.
  • Non-toxic and chemically inert to biological fluids and most acids
  • Easy to grip with gloves
  • Temperature Range: –50 to 90 °C
  • Can be gas (EtO) sterilized
Designed for measurements, these graduated transfer pipettes feature molded-in graduation marks that will not fade or wash off.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty