Would you like to log in again?
G-SEP™ DEAE Agrose Fast Flow, G-Biosciences
Supplier: G-Biosciences
G-SEP™
786-968 786-967
76285-974EA 1327.67 USD InStock
76285-974 76285-972
G-SEP™ DEAE Agrose Fast Flow, G-Biosciences
Protein Purification Protein Purification Resins
G-Sep™ DEAE Agarose Fast Flow (FF) resin is a weak anion exchanger composed of highly cross-linked 6% agarose beads, with diethylaminoethyl (DEAE) weak anion exchange groups.
  • Used for ion exchange chromatography
  • Matrix: Cross-linked agarose beads, 6%
  • Ligand: Diethylaminoethyl
The resin is extensively used for biomacromolecule separations in both research and industrial applications. The G-Sep™ DEAE Agarose Fast Flow (FF) resins have high chemical stability, allowing well proven cleaning-in-place (CIP) and sanitization protocols and have been the industrial standard for ion exchange chromatography during recent decades.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty