Would you like to log in again?
Experimentation Kits, Dot Blot Analysis
Supplier: G-Biosciences
BE-502
76286-340EA 179.35 USD InStock
76286-340
Experimentation Kits, Dot Blot Analysis
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Dot blotting is a simple technique to identify a known protein in a biological sample. The ease and simplicity of the technique makes dot blotting an ideal diagnostic tool.
  • Simulated test samples
  • Teaches immunodetection technique
  • Hands-on dot blot analysis technique with necessary reagents and supplies
The method involves applying small volumes of antigens or test samples on protein binding membrane; the membrane captures and immobilizes proteins. The proteins immobilized on the membrane are probed with a specific antibody and a matching specific detection reagent. In this lab activity students screen test samples from a population and identify those who are carriers of a disease. Students learn how dot blotting applied in the real world, for example in clinical testing, forensic, and research laboratories.
Delivery information: Supplied with components needed for hands-on experimentation for six workstations of 4-5 students or 24-30 students. Supplied with teacher’s guide and separate student’s guides.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty