Would you like to log in again?
FREEZECELL HEX SHAPE F/12VIALS 1ML/2ML
Supplier: CRYSTAL TECHNOLOGY & INDUSTRIES
Freezecell Hex Shape F/12Vials 1Ml/2Ml^ Freezecell, Hex Shape, Holds 12 Vials 1ml/2ml, Easy to use, Low cost, Minimal, and wont use up a lot of freezer space, Cooling rate consistency is very good, freeze an individual experimental, or a full load, Transfer the samples conveniently, No pre-cooling
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FREEZECELL HEX SHAPE F/12VIALS 1ML/2ML 76287-388 FZC-01 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty