Would you like to log in again?
Vaplock™ Chemical Exhaust Filter Adapters, Cole-Parmer
Supplier: Cole Parmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
12019-88 12019-97 12019-86 12019-94 12019-89 12019-90 12019-99 12019-98 12019-92 12019-85 12019-87 12020-00 12019-96 12019-93 12019-91
76287-672EA 63.79 USD InStock
76287-672 76287-486 76287-494 76287-670 76287-484 76287-492 76287-482 76287-490 76287-480 76287-676 76287-666 76287-710 76287-674 76287-488 76287-664
Vaplock™ Chemical Exhaust Filter Adapters, Cole-Parmer
Fittings and Connectors
Exhaust filters may be attached to various point-source emissions by the use of Vaplock™ adapters.
Filter Adapters offer versatile configurations.
Please note that options for justrite safety containers are made to fit either stainless steel or polypropylene quick disconnects, but are not interchangeable.
Vaplock™ EF-100 and EF-200 exhaust filters may be attached to various point-source emissions by use of the appropriate adapter. Each filter includes a 1/4" female NPT port; select from the list below to adapt filters to a typical system, or inquire for assistance with special requirements. All adapters are reusable.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty