Would you like to log in again?
Vaplock™ GL 38 Supply Caps, Cole-Parmer
Supplier: Cole Parmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
12018-23 12018-13 12018-12 12018-20 12018-17 12018-19 12018-16 12018-14 12018-21 12018-18 12018-22 12018-15
76287-718EA 61.64 USD InStock
76287-718 76287-726 76287-716 76287-724 76287-944 76287-516 76287-942 76287-722 76287-514 76287-720 76287-512 76287-510
Vaplock™ GL 38 Supply Caps, Cole-Parmer
Closures Safety Caps and Safety Waste Caps
The Vaplock™ closed system design reduces chemical evaporation and exposure to hazardous vapors with Vaplock™ closed system solvent reservoir caps.
  • Anti-twist design prevents twisting of tubing when the cap is attached or removed as the inner body rotates independently of the outer collar
  • Chemical compatibility with laboratory solvents
Durable port threads are designed to accept industry-standard 1/4-28 threaded fittings. Some caps include robust stainless steel thread inserts to minimize cross-threading and damage to soft PTFE ports. For use with semi-rigid tubing equal to or less than 1/8" (3.2 mm) O.D.
Chemically compatible caps are constructed of inert PTFE. The body of the cap provides ideal chemical compatibility with laboratory solvents. Under normal use, the outer PP collar is non-wetted. Caps may be autoclaved to 140 °C. Combine with secure fluid connections and Vaplock air inlet or relief valves to maintain pressure equilibrium within the bottle.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty