Would you like to log in again?
Vaplock™ DIN 50 Waste Caps and Waste Kits, Cole-Parmer
Supplier: Cole Parmer
12018-90 12018-91 12018-86 12018-88 12018-89 12018-87
76287-794KT 445 USD InStock
76287-794 76287-792 76287-802 76287-800 76287-798 76287-796
Vaplock™ DIN 50 Waste Caps and Waste Kits, Cole-Parmer
Closures Safety Caps and Safety Waste Caps
The Vaplock™ closed system design reduces chemical evaporation and exposure to hazardous vapors.
  • Minimize exposure to harmful chemical vapors
  • Collect solvent waste from multiple effluent streams
  • Highly adaptable and conformable to a broad range of containment types
Featuring a vapor tight connection, waste manifolds are designed to fit standard DIN 50 threaded lab bottles. Chemical-resistant materials of construction include polypropylene or PTFE manifold and insert, with polypropylene spin collar. These versatile manifolds can be configured for barbed connections, compression fittings, 1/4-28 fittings, and ferrule sets. All manifolds are designed to accommodate the EF-100 Filter and indicator.
Ordering information: Kits include (1) 10-port waste manifold, (3) 1/4" NPT polypropylene plugs, (6) 1/4-28 PTFE plugs, (1) 1/2" NPT polypropylene plug, (1) organic vapor filter, (2) 2.2 mm O.D. hard wall tubing connections, (1) 2.5 mm O.D. hard wall tubing connection, (3) 1/8" O.D. hard wall tubing connections, (2) 1/16" O.D. hard wall tubing connections, (3) barbed tubing connection 1/4" I.D. tubing×1/4" NPT polypropylene, (1) barbed tubing connection 1/8" I.D. tubing×1/4" NPT polypropylene, (1) barbed tubing connection 3/16" I.D. tubing×1/4" NPT polypropylene, (1) barbed tubing connection 3/8" I.D. tubing×1/4" NPT polypropylene, (1) barb to barb Y connector with 1/4" (6.25 mm) barbs, polypropylene, (1) hard wall tube extension with union connections and 5' of 1/8" O.D. PTFE tubing, (1) filter adapter 1/4" NPT×1/2" NPT polypropylene, and (1) breakthrough color indicator for Vaplock™ organic vapor filter.
Container not included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty