Would you like to log in again?
Vaplock™ Large-Bore Tubing Connections, Cole-Parmer
Supplier: Cole Parmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
12020-41 12020-37 12020-36 12020-43 12020-39 12020-38 12020-42 12020-40
76287-928PK 17.97 USD InStock
76287-928 76287-926 76287-938 76287-924 76287-936 76287-934 76287-932 76287-930
Vaplock™ Large-Bore Tubing Connections, Cole-Parmer
Fittings and Connectors
For liquid-tight connection of semi-rigid tubing (1/4" to 3/4" O.D.) to NPT ports.
Large-bore tubing connections for 1/4" NPT and 1/2" NPT ports.
Wrap NPT threads 3 to 4 times with PTFE tape and wrench tighten for leak-resistant connections.
These compression fittings designed for liquid-tight connection of semi-rigid tubing to NPT ports, for 1/4" to 3/4" O.D. tubing. Useful for pressurized applications under 100 psi. Select based on tubing outer diameter. All fittings are comprised of two-piece molded polypropylene components and do not include a separate ferrule.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty