Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vaplock™ Wako Supply Caps, 38 mm, Cole-Parmer

Supplier: Cole Parmer

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

12018-39 12018-44 12018-46 12018-45 12018-37 12018-43 12018-36 12018-42 12018-40 12018-38 12018-47 12018-41
76287-948EA 66.52 USD InStock
76287-948 76287-728 76287-946 76287-738 76287-746 76287-736 76287-744 76287-734 76287-742 76287-732 76287-740 76287-730
Vaplock™ Wako Supply Caps, 38 mm, Cole-Parmer
Closures Safety Caps and Safety Waste Caps
The Vaplock™ closed system design reduces chemical evaporation and exposure to hazardous vapors with Vaplock™ closed system solvent reservoir caps.

  • Anti-Twist design prevents twisting of tubing when the cap is attached or removed
  • Chemical compatibility with laboratory solvents

Durable port threads are designed to accept industry-standard 1/4-28 threaded fittings. Some caps include robust stainless steel thread inserts to minimize cross-threading and damage to soft PTFE ports. For use with semi-rigid tubing equal to or less than 1/8" (3.2 mm) O.D. The anti-twist design prevents twisting of tubing when the cap is removed or attached, as the inner body rotates independently of the outer collar.

Chemically compatible caps are constructed of inert PTFE. The body of the cap provides ideal chemical compatibility with laboratory solvents. Under normal use, the outer PP collar is non-wetted. Caps may be autoclaved to 140 °C. Combine with secure fluid connections and Vaplock air inlet or relief valves to maintain pressure equilibrium within the bottle.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty