Would you like to log in again?
Wheaton® MicroLiter® ULTRAPure Septa Crimp Caps and Screw Caps, DWK Life Sciences
Supplier: DWK Life Sciences
MicroLiter
09-0039N 11-0038A 11-0039A 09-0038N
76045-382CS 48.67 USD InStock
76045-382 76045-380 76288-112 76288-114
Wheaton® MicroLiter® ULTRAPure Septa Crimp Caps and Screw Caps, DWK Life Sciences
Closures Chromatography Vial Closures
Caps are manufactured from the highest purity, ultra-low bleed silicone available in the market.
  • 9 mm natural polypropylene caps or 11 mm silver aluminum crimp caps
  • Available in non-slit and pre-slit formats
  • Ultra-low bleed blue/teal silicone Shore A is laminated to white PTFE film, 3 mm thick
  • Silver aluminum crimp caps are lacquer coated
Certified ULTRAPure septa is manufactured from the highest purity, ultra-low bleed silicone available in order to combat the leachable effects of silicone without trading off the ability to be pierced multiple times. The MicroLiter certified pure septa provides accurate and reproducible LC-MS and GC-MS results.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty