Would you like to log in again?
Sample and Storage Tubes, Globe Scientific
Supplier: Globe Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6030 6038 6059 6034 6039 6053 6052 6035 6032 6054 6033 6055 6051 6031 6050 6058
10028-098CS 309.81 USD InStock
10028-098 10028-100 10028-102 10028-104 89209-790 10028-180 10028-182 10028-184 10028-096 76288-142 76288-144 76288-140 76288-138 76288-134 76288-146 76288-136
Sample and Storage Tubes, Globe Scientific
Tubes Test Tubes
Sample and storage tubes and caps have been designed for the storage and transportation of biological material.
  • Manufactured from non-toxic polypropylene
  • External threads provide a smooth and uniform inner surface
Manufactured from polypropylene for strength and clarity. The vial has external threads, which provide a smooth and uniform inner surface to reduce the risk of contamination. The screw caps are also produced from polypropylene, providing the same coefficient of expansion as the tubes. This, along with a polypropylene inner lip, ensures a leak-proof seal, even at changing temperatures. The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn.
The caps are available with or without a silicone O-ring, and are available in a variety of colors.
Ordering information: All screw caps are sold separately.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty