Would you like to log in again?
Potato Flake Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
W59 L48
75993-540PK 41.4 USD InStock
75993-540 76288-258
Potato Flake Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Hardy Diagnostics Potato Flake Media is used in the cultivation, isolation, and induction of sporulation of fungi from clinical specimens.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty