Would you like to log in again?
Chromogenic Western Blot Kit (Anti-Mouse), Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT182-0001
76288-290EA 672 USD InStock
76288-290
Chromogenic Western Blot Kit (Anti-Mouse), Enzo Life Sciences
Assays Immunological Assays
Chromogenic Western Blot kit with increased signal and sensitivity.
  • More sensitive detection - able to detect lower concentrations of protein
  • Easy to use - all reagents are ready-to-use or ready-to-dilute, saving precious time
  • Easily adaptable - works with a wide range of mouse-derived proteins
The Chromogenic Western Blot Kit (anti-mouse) can be used to identify primary antibodies on a Western Blot using immunodetection. The kit contains a complete reagent set, with optimized ready-to-use or ready-to-dilute reagents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty