Would you like to log in again?
Dopamine ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT188-0001
76288-304EA 910 USD InStock
76288-304
Dopamine ELISA Kit, Enzo Life Sciences
Assays ELISAs
Sensitive ELISA kit for the quantification of Dopamine in human samples.
  • High-sensitivity ELISA enables quantification of low amounts of Dopamine
  • Rapid quantitative results in <2 hours
  • High-throughput immunoassay measures up to 40 samples in duplicate per 96 well kit
  • Pre-coated plates reduce errors and save time
It is used for the quantitative determination of human dopamine in serum, plasma, tissue homogenates, and other biological fluids.
The Dopamine ELISA kit is a colorimetric immunoassay kit with results in <2 hours. Absorbance is read at 450 nm. Measure up to 40 samples in duplicate per 96 well  kit.
Delivery information: Contains Microtiter plate, Lyophilized Standard, Sample/standard dilution buffer, Concentrated biotin detector antibody, Antibody Dilution Buffer, HRP-Streptavidin Conjugate, Conjugate Dilution Buffer, TMB Substrate, Stop Solution, Wash Buffer (25X), Plate Sealer.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty