Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Wickerham Card, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z08
76288-330EA 14.99 USD InStock
76288-330
Wickerham Card, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
Designed for turbidity adjustments to a McFarland standard.
A plastic laminated card with thick black and white lines to facilitate the preparation of bacterial and yeast suspensions when comparing the turbidity to a McFarland Standard. A handy visual aid to be used for the preparation of inocula for the disk diffusion and other tests that require a specified McFarland turbidity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty