Would you like to log in again?
Mouse Recombinant FGF basic
Supplier: Enzo Life Sciences
201-824-0100 201-824-0010 201-824-0050
76001-952EA 321.81 USD InStock
76001-952 76001-950 76288-346
Mouse Recombinant FGF basic
Proteins and Peptides
Produced in E. coli. Non-glycosylated protein, containing 145 amino acids.
  • Carrier-free
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty