Would you like to log in again?
CellExp™ Human Recombinant ErbB3 (from HEK293 cells)
Supplier: BioVision
CellExp™
AB285846-250UG
76289-064EA 5791.6 USD InStock
76289-064
CellExp™ Human Recombinant ErbB3 (from HEK293 cells)
Proteins and Peptides
ErbB3, also called human epidermal growth factor receptor 3 (HER3), is a type I membrane glycoprotein and a member of the ErbB family. ErbB family members serve as receptors for the epidermal growth factors (EGF). ErbB3 is expressed in normal adult human gastrointestinal tract, reproductive system, skin, nervous system, urinary tract, and endocrine system. ErbB3 is the only member with a defective kinase domain among ErbB family members. However, ErbB3 can form heterodimers with ErbB2 to complement a high affinity receptor complex. ErbB3 has been shown to bind the ligands heregulin and NRG-2. The lethal congenital contracture syndrome 2 (LCCS2) disease is caused by mutations affecting ErbB3. ErbB3 null mouse embryos show severely underdeveloped atrioventricular valves, leading to death at embryonic day 13.5.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty