DYNEO™ DD refrigerated/heating circulator combinations with stainless steel bath tanks for internal or external temperature applications from –20 to 200 to –50 to 200 °C.
- Infinitely adjustable pressure pump
- Easy switching between internal and external circulation
- Large color TFT display, multilingual interface
- Integrated programmer
- PT100 port facilitates external temperature control via PT100 sensor
- USB port
- Optional RS232 interface or analog interfaces
For internal and external temperature applications. High quality stainless steel bath tanks with bath lid and drain tap.
DYNEO DD circulator features a color TFT display with central knob controller for operating/programming. Integrated programmer. A lever below the display enables simple adjustment between internal and external fluid flow. The USB port facilitates external communication with models options to add RS232 interface or analog interface. Operation in °C or °F. Pump connections: M16×1 male threads.
Certifications: CE, UKCA. DIN 12876-1 Class III.
Ordering information: Each unit is supplied with a pair of 8 and 12 mm hose barbs. Fluid, adapters, tubing sold separately.
Delivery information: Includes DYNEO DD immersion circulator, refrigeration unit, mounting bridge, bath tank lid, pair of 8 and 12 mm hose barbs, integrated drain tap.