Would you like to log in again?
Conical Tubes with Screw Caps
Supplier: Eppendorf
0030122526 0030122410 0030122151 0030122178 0030113497 0030122534 0030113489 0030122500 0030122194 0030113489 0030122160 0030122429 0030122526 0030122445 0030122534 0030122186 0030122542 0030113497 0030122445 0030122259 0030122224 0030122542 0030122267 0030122437
76338-501BG 15.45 USD InStock
76338-501 CA76683-152 76683-152 CA77391-924 76587-090 CA77391-922 77391-924 76019-022 77391-922 76019-024 75875-238 75874-294 76289-502 76289-500 76200-146 CA76587-090 76587-092 CA76587-092 76200-144 76587-094 76338-492 CA76587-094 76338-498 76338-496
Conical Tubes with Screw Caps
Tubes Microcentrifuge Tubes
Eppendorf® conical tubes with screw caps offer reliable sample containment and leak-free storage. SafeCode variants enhance sample identification, while the BioBased options, manufactured using renewable resources, provide an eco-friendly alternative without compromising performance.
  • Polypropylene (PP) tubes, High-Density Polyethylene (HDPE) screw caps
  • Ensures leak-free storage and protects sample integrity
  • Suited for a wide range of laboratory tasks, enhancing flexibility
  • High-quality materials for long-lasting, robust performance
  • Clear, visible graduations for quick volume assessment
  • Facilitates optimal sample recovery and efficient centrifugation
  • Available in 15, 25, and 50 ml formats
  • BioBased varients aid in the reduction of consumable-related carbon footprint in the lab
  • 2D Datamatrix (SafeCode) option features 2-level coding for safe sample identification
Eppendorf® Conical Tubes with Screw Caps epitomize secure sample containment. The screw cap ensures leak-free storage, safeguarding sample integrity during transportation and storage. Versatile for diverse lab tasks, these tubes offer durability and reliable performance. Clear graduations facilitate easy volume assessment, while the conical design aids optimal sample recovery and efficient centrifugation.
BioBased option: The Eppendorf Tubes® BioBased option open up new doors to make laboratory work significantly more sustainable without jeopardizing experimental results. By using a second-generation biobased polymer made from 90% renewable resources (e.g. waste and residues from vegetable oil refining or waste cooking oil) the product-related carbon footprint of these tubes is significantly reduced. BioBased varients aid in the reduction of consumable-related carbon footprint in the lab. The bio-based tubes have identical material properties and technical specifications as non-BioBased tubes.
2D SafeCode Options (Barcoded): A clean and stable label on the side or on the top of the vessel can make scientific life so much easier. Eppendorf tubes are available with SafeCode, a well-designed 2D barcoding system. Take advantage of the pre-labeled and durable 2-level coding of the SafeCode system (datamatrix code and clear coding) to enable safe sample identification. The safe sample ID is supported by additional online information about the dedicated consumable at the Eppendorf dataport.
A barcode is an optical representation of data which is machine-readable. Classic 1D barcodes code data by varying the width of the parallel lines as well as their spacings. Two-dimensional codes (2D) are based on dots, hexagons, rectangles, or other geometric patterns. These codes are also called data matrix codes. ECC200 error correction function enables safe code reading with up to 25% damaged codes.
Manufactured without the use of slip agents, plasticizers, and biocides - substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
Certifications: ACT label (25 ml), lot-specific certificates for the purity grades PCR clean, sterile; ISCC PLUS declarations (BioBased varient) can be downloaded from the Eppendorf website.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty