Would you like to log in again?
Eppendorf FAST PCR Tube Strips
Supplier: Eppendorf
Eppendorf FAST
0030124910 0030124928 0030124901
76289-506EA 371.25 USD InStock
76289-506 76289-504 76289-508
Eppendorf FAST PCR Tube Strips
Tubes PCR Tubes
Eppendorf Fast PCR tube strips made of polyethylene transfer heat faster than tubes made of traditional plastics. The fast PCR tube strips are particularly suited to scientists using fast PCR reagent kits, scientists trying to speed up protocols or cyclers with high heating and cooling rates.
  • Ideal for use with fast cyclers and fast enzymes
  • Eight reaction tubes in strip-format – ideal for small sample volumes
  • Easily adaptable for automation
  • Sealable using flat or domed strip lids
  • Certified free from human DNA, DNase, RNase and PCR inhibitors
The fast PCR tube strip allows scientists using fast cyclers (such as the Eppendorf Mastercycler X50) and fast enzymes to obtain higher PCR yield. This is achieved by the faster heat transfer from block to reaction, allowing for better control over the reaction. The tube strips are suitable for fast end-point PCR and real-time PCR applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty