Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DURAN® GL 45 Bromobutyl Rubber Stoppers, DWK Life Sciences

Supplier: DWK Life Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

DURAN®
292062803
76289-760CS 29.21 USD InStock
76289-760
DURAN® GL 45 Bromobutyl Rubber Stoppers, DWK Life Sciences
Closures Test/Sample Tube Closures
DURAN® Bromobutyl Rubber closures provide a gas tight seal for all GL45 laboratory bottles with sizes from 100 to 20000 ml. Bromobutyl rubber is essentially impermeable to most gases and provides a controlled environment inside the glass bottle for oxygen sensitive materials. Useful for maintaining anaerobic culture conditions. Butyl rubber allows for multiple punctures providing easy access to the contents with a syringe.

  • Low permeability to air, gases and moisture
  • Material is virtually free of organics
  • Elastic material for effective sealing and reliable self-sealing after needle piercing
  • Upper surface features seven numbered hemispherical impressions for needle-piercing
  • Wide usable temperature range: –50 to +121 °C

They are used in anaerobic culture of microorganisms, storage of anaerobic culture media, aerobic methane oxidation, storage of oxygen sensitive materials and applications requiring gas purging or sample transfer via a syringe.

These stoppers are suitable for autoclaving or steam sterilizing at 121 °C for 20 minutes.

Used with aperture screw cap for extra security.

Certifications: Conforms to EP 3.2.9 Type I closure for chemical and penetration tests.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty