Would you like to log in again?
93-360 Thinnest, Chemical Resistant Disposable Cleanroom Gloves, Microflex®, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
93360100 93360110 93360090 93360080 93360060 93360070
76289-858CS 510.32 USD InStock
76289-858 76289-856 76289-850 76289-860 76289-854 76289-852
93-360 Thinnest, Chemical Resistant Disposable Cleanroom Gloves, Microflex®, Ansell
Gloves
This is made of an innovative, synthetic composite material for tough chemical protection.
  • Three layer design for superior protection
  • Thin construction
  • Easy donning
  • Non-particulating packaging
The extra soft formulation and ergonomic design provide a comfortable fit and feel for extended wear times.
Certifications: CE 0086 marked. EN ISO 374-1 Type A: JKLOPST chemical hazards. EN ISO 374-5: VIRUS. EN 388: 2000X mechanical hazards. EAC TP to TC 0192011. EN 421.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty