Would you like to log in again?
Aven ProVue Solas Magnifying Lamp, Aven
Supplier: Aven
ProVue Solas
26501-XL35
76289-866EA 252.84 USD InStock
76289-866
Aven ProVue Solas Magnifying Lamp, Aven
Magnifiers
Offers flexibility of using the 5-Diopter Lens (2.25× magnification).
  • 60 ultra bright SMD LEDs with brightness controls
  • Fully enclosed spring balanced arm
  • Protective lens cover
  • Heavy duty table clamp
Optional 3-Diopter Lens (1.75× magnification, sold separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty