Would you like to log in again?
TIP CLIPTIP ST OR 30-300UL/8X96 RACK LR
Supplier: Thermo Fisher Scientific
Tip Cliptip St Or 30-300Ul/8X96 Rack Lr^ Pipette tips, ClipTip*, Length: 100mm, Sterile, Colour Coded: Orange, For use with E1-ClipTip and F1-ClipTip pipettes, utilises low retention technology to enable maximium sample recovery, free of RNase, ATP and endotoxin contamination, Volume: 30-300ul
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TIP CLIPTIP ST OR 30-300UL/8X96 RACK LR 76289-882 94410613 Pack of 768 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty