Would you like to log in again?
TIP CLIPTIP STERILE 10X96/INSERT 2/RK LR
Supplier: Thermo Fisher Scientific
Tip Cliptip Sterile 10X96/Insert 2/Rk Lr^ Pipette tips, ClipTip*, Length: 100mm, Sterile, Colour Coded: Orange, For use with E1-ClipTip and F1-ClipTip pipettes, utilises low retention technology to enable maximium sample recovery, free of RNase, ATP, Volume: 30-300ul, 10 reload inserts, 96 each
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TIP CLIPTIP STERILE 10X96/INSERT 2/RK LR 76289-886 94410618 Pack of 960 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty