Would you like to log in again?
ClipTip® Pipette Tips, 12.5, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ClipTip™
94420043 94410040 94410063 94410043 94410060 94420063
76289-900PK 129.02 USD InStock
76289-900 10030-778 76289-894 76289-892 10030-780 10030-782
ClipTip® Pipette Tips, 12.5, Thermo Scientific
Pipette Tips
Thermo Scientific ClipTip® 12.5 pipette tips feature a unique ‘Snap and Lip’ seal technology to attach and seal the tips on each channel with minimal tip attachment and ejection force.
  • Certified free of RNase, DNase, DNA, ATP, and pyrogen contamination
  • Available in nonsterile, sterile, and filtered formats
  • Each tip insert has ink-jetted lot and volume information
Convenient color keys make it easy to match the color of the E1-ClipTip® pipettor with the corresponding ClipTip® pipette tip rack insert.
ClipTip® 12.5 pipette tips are suitable for pipetting 0.5 to 12.5 µl volumes with the E1-ClipTip® electronic pipettors.
Certifications: Manufactured according to ISO 9001 and ISO 13485.
Ordering information: A lot specific Certificate of Conformity is provided with each unit box.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty