Would you like to log in again?
TIP CLIPTIP STERILE 0.5-12.5UL/384 TIP
Supplier: Thermo Fisher Scientific
Tip Cliptip Sterile 0.5-12.5Ul/384 Tip^ Pipette tips, ClipTip*, Length: 41 mm, Colour Coded: Pink, Sterile, For use E1-ClipTip pipettes, utilises low retention technology to enable maximium sample recovery, free of RNase, DNase, DNA, ATP and endotoxin contamination, Volume: 0.5-12.5 ul, 384 tips
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TIP CLIPTIP STERILE 0.5-12.5UL/384 TIP 76289-898 94410073 Pack of 3,840 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty