Would you like to log in again?
DuPont™ ProClean® 2 Heavy Duty Shoe Covers with Traction
Supplier: DuPont
ProClean®
GB440SBU2XMP GB440SBUXL0B GB440SBU000B GB440SBU2X0B
76290-240CS 134.51 USD InStock
76290-240 76290-242 76290-244 76290-246
DuPont™ ProClean® 2 Heavy Duty Shoe Covers with Traction
Disposable Garments
DuPont™ ProClean® 2 shoe covers are made from spunbond meltblown spunbond (SMS) with a traction sole.
  • SMS shoe covers
  • Serged seams
  • Elastic opening
  • Heavy duty material
Shoe covers feature a traction sole with serged seams and an elastic opening. Multiple packaging options are available.
Certifications: Thickness meets ASTM D3776.
Packaging: Shoe covers in Multi Pack option are packaged in a multi-pack dispenser with 400 per case. Shoe covers in Bulk Packed option are bulk packaged and placed in transparent poly liners with 500 covers per case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty