Would you like to log in again?
Short-Path Still Distillation Head, Ace Glass
Supplier: Ace Glass
6611-30 6611-20 6611-10
76214-412EA 2116.43 USD InStock
76214-412 76266-078 76291-838
Short-Path Still Distillation Head, Ace Glass
Distillation Components Distillation Heads
Short-Path distillation head with integral vigreux vapor path column.
  • Distillation head made of high-quality borosilicate glass
  • Head has Ace-Thred condenser inlet/outlets, and vacuum monitoring port
  • Can be used with Ace-Safe hose connections
  • Condenser length given is the effective length
Also has drip tip distribution receiver joint and a 14/20 thermometer/probe port.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty