Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sapphire Biomolecular Imager, Azure Biosystems
Supplier: Azure Biosystems
IS1028 IS1023 IS2012 IS1004 IS2009 IS1018 IS1025 IS1024 IS1027 IS1003 IS1026 IS2011
76317-608EA 125596.22 USD InStock
76317-608 76291-952 76318-250 76317-606 76291-950 76318-252 76318-254 76317-610 76318-256 76318-248 76317-604 76291-948
Sapphire Biomolecular Imager, Azure Biosystems
Photodocumentation Systems
The next generation lab imaging platform.
  • With most imaging systems a decision between application flexibility and image quality has to be made
  • Next generation laser scanning system which offers the best of both worlds
  • Unparalleled flexibility with excellent sensitivity and image quality
  • Capture software will deliver high quality quantifiable data
Up to four solid state lasers (488, 520, 658 and 784 nm) offering ultimate excitation sensitivity. Photon multiplier tube (PMT) for fluorescence and phosphor imaging, avalanche photodiodies (APD) for near-infrared imaging and a CCD sensor for chemiluminescent and visible imaging. Ultra-wide dynamic range for imaging and quantifying low and high abundance samples simultaneously. Image resolution down to 10 microns for high-quality image analysis.
Order Now
ORDER
ACCESSORIES
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty