Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ao Absorbance Microplate Reader, Azure Biosystems
Supplier: Azure Biosystems
AC3126 AC3102 AC3104 AC3103 AC3120 AC3108 AC3121 AC3200 AC3107 AC3127 AC3105 AC3112 AC3118 AC3124 AC3116 AC3122 AC3100 AC3201 AC3128 AC3101 AC3117 AC3111 AC3000 AC3106 AC3113 AC3109 AC3115 AC3114 AC3119 AC3125 AC3110 AC3123
76291-988EA 224.83 USD InStock
76291-988 76291-986 76291-982 76292-030 76292-010 76292-032 76292-038 76292-016 76292-018 76292-012 76292-034 76292-036 76292-014 76291-998 76291-996 76291-994 76291-992 76292-042 76292-020 76292-044 76292-000 76292-022 76292-040 76292-028 76292-006 76292-008 76292-024 76292-002 76292-046 76292-004 76292-026 76291-990
Ao Absorbance Microplate Reader, Azure Biosystems
Microplate Readers
96 well plate absorbance microplate reader.
  • Offers a simple user interface
  • Absorbance-based reading offers maximum sensitivity
  • Best read speeds for kinetic assays
  • High flexibility
  • Lower cost compared to fluorescent and luminescent-based readers
Its top-reading design is ideal for solution-based assays such as ELISAs, DNA and protein quantification, and enzyme kinetics.
Stand-alone, top absorbance reader. Large, easy-to-use 7" touch screen interface. Pre-set and custom protocols. Single and dual wavelength detection modes. 8 position filter wheel: 5 standard filters (405, 450, 492, 595, and 630) plus 3 options from 340 to 750 nm. Built-in shaker with speed selection. Motorized door for easy sample loading.
Order Now
ORDER
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty